Learn More
CD45 Mouse anti-Human, BB515, Clone: HI30, BD Horizon™
Mouse Monoclonal Antibody
Marke:  BD Biosciences 564586
Additional Details : Gewicht : 0.01000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15833739
  • 157.00 CHF / 1 Stück
Verfügbar ab
01-06-2021
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow cytometry.
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow cytometry.
Spezifikation
Spezifikation
CD45
HI30
Vol. Per test: 5μL
Purified
Murine
Mouse IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Monoclonal
Flow Cytometry
BB515
Aqueous buffered solution containing ≤0.09% sodium azide.
PTPRC; CD45R; LCA; L-CA; Leukocyte Common Antigen; B220; T200; GP180; LY5
Human Peripheral Blood Leucocytes
25 Tests
Cell Biology