missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Monoclonal Antibody (HI30), APC-Alexa Fluor™ 750, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MHCD4527
Additional Details : Gewicht : 0.50000kg
Beschreibung
The allophycocyanin (APC)-Alexa Fluor® 750 dye tandem conjugate allows for multicolor measurements using excitation by lasers that emit from 633 to 650 nm.
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD45 | |
Monoclonal | |
APC-Alexa Fluor 750 | |
PTPRC | |
B220; cd45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; leukocyte common antigen, CD45; loc; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase lambda; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T | |
Mouse | |
purified | |
ASR | |
5788 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
HI30 | |
PBS with BSA, sucrose and 0.1% sodium azide | |
P08575 | |
PTPRC | |
CD45 antigen | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |