missing translation for 'onlineSavingsMsg'
Learn More

CD45.1 Mouse, FITC, Clone: A20, enQuireBio™

Mouse Monoclonal Antibody

115.50 CHF - 284.55 CHF

Specifications

Antigen CD45.1
Klon A20
Konzentration 0.5mg/mL
Anwendungen Flow Cytometry
Klassifikation Monoclonal
View More Specs

Products 3
Product Code Brand Menge Price Quantity & Availability  
Product Code Brand Menge Price Quantity & Availability  
15982008
View Documents
EnquireBio
QAB42-F-25ug
25 μg
115.50 CHF
25 Mikrogramm
View alternative products
Save Up to 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
15972008
View Documents
EnquireBio
QAB42-F-100ug
100 μg
220.50 CHF
100 Mikrogramm
View alternative products
Save Up to 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
15992008
View Documents
EnquireBio
QAB42-F-500ug
500 μg
284.55 CHF
500 Mikrogramm
View alternative products
Save Up to 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Description

Description

Specifications

Specifications

CD45.1
0.5mg/mL
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
19264
IgG2a κ
Protein A/G Purified
A20
Flow Cytometry
FITC
RUO
NP_001255215
Ptprc
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.