Learn More
CD45.1 Mouse anti-Mouse, PE, Clone: A20, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech PE-65092-100UG
Beschreibung
The CD45.1 antibody from Proteintech is a mouse monoclonal antibody generated with sjl mouse thymocytes and splenocytes of mouse CD45.1. This antibody recognizes mouse antigen. The CD45.1 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Spezifikation
CD45.1 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
Primary | |
Mouse |
Flow Cytometry | |
A20 | |
PE | |
100 μg | |
19264 | |
IgG2a |
Produktvorschläge
Customers who viewed this item also viewed.