Learn More
CD44std Monoclonal Antibody (SFF-2), Biotin, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marke: Affymetrix eBioscience BMS113BT
Additional Details : Gewicht : 0.23750kg
Beschreibung
Description: BMS113 specifically recognizes human standard CD44, but cannot be used to distinguish between splice variants (exon v3-v10). CD44 is a cell membrane associated, polymorphic glycoprotein with apparent molecular weights ranging from 85 kDa to 250 kDa. CD44 isoforms participate in a wide variety of cell-cell or cell-matrix interactions including lymphocyte homing, establishment of B and T cell immune responses, tumor metastases formation and inflammation. Three isoform categories of the CD44 molecule have been identified: 1) an 80-90 kDa isoform, the so-called standard form named CD44std, which is widely distributed on several hematopoietic and nonhemato-poietic cells including all subsets of leukocytes, monocytes, erythrocytes, many types of epithelium, mesenchymal elements like fibroblasts, smooth muscle cells and glial cells of the central nervous system, 2) a medium size category of 110-160 kDa which is weakly expressed on epithelial cells and highly expressed in some carcinomas and 3) a category which includes very large isoforms of 250 kDa covalently modified by the addition of chondroitin sulfate. These bigger isoforms of CD44 arise by alternative splicing of one or more ""variant"" exons (v2-v10) into the extracellular part of the 90kDa constant form molecule. Compared to the standard CD44, all larger isoforms are expressed in a much more restricted fashion, only in a few normal tissues or on the surface of certain tumor cells.
CD44std Monoclonal antibody specifically detects CD44std in Human samples. It is validated for Flow Cytometry.Spezifikation
CD44std | |
Monoclonal | |
10 μL/Test | |
TBS with 1% BSA and 0.02% sodium azide; pH 7.2-7.4 | |
P16070 | |
CD44 | |
Affinity chromatography | |
RUO | |
960 | |
2-8°C | |
Liquid |
Flow Cytometry | |
SFF-2 | |
Biotin | |
CD44 | |
AU023126; AW121933; AW146109; Bovine CD44; CD44; CD44 antigen; CD44 antigen-like protein; CD44 molecule; CD44 molecule (Indian blood group); CD44 protein; CD44 variant; CD44A; CD44-like protein; CDW44; cell surface glycoprotein CD44; Cell surface glycoprotein CD44 (hyaluronate binding protein); cell-surface glycoprotein; chondroitin sulfate proteoglycan 8; CSPG8; ECMRIII; ECMR-III; Epican; extracellular matrix receptor III; extracellular matrix receptor-III; GP90 lymphocyte homing/adhesion receptor; HCELL; hematopoietic cell E- and L-selectin ligand; heparan sulfate proteoglycan; HERMES; Hermes antigen; homing function and Indian blood group system; HUTCH-I; hyaluronan receptor; Hyaluronate receptor; IN; LHR; Ly-24; lymphocyte antigen 24; lymphocyte surface antigen precursor CD44; MC56; MDU2; MDU3; METAA; MIC4; PGP I; Pgp1; PGP-1; PGPI; PGP-I; Phagocytic glycoprotein 1; phagocytic glycoprotein I; putative Bos taurus CD44; RHAMM; sCD 44; sCD44; soluble CD 44; soluble CD44 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.