Learn More
CD43 Rat anti-Mouse, PE, Clone: S7, BD
Rat Monoclonal Antibody
154.00 CHF - 520.00 CHF
Spezifikation
Antigen | CD43 |
---|---|
Klon | S7 |
Konzentration | 0.2mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15821409
|
BD Biosciences
561857 |
25 μg |
154.00 CHF
1 Stück |
Verfügbar ab: 22-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15827118
|
BD Biosciences
553271 |
0.2 mg |
520.00 CHF
1 Stück |
Verfügbar ab: 22-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The S7 monoclonal antibody specifically binds to the 115 kDa glycosylated form of CD43 (Ly-48, Leukosialin). CD43 is expressed on IL-7-responsive pro-B cells, plasma cells, peritoneal and splenic CD5+ B cells (B-1 cells), granulocytes, monocytes, macrophages, platelets, natural killer cells, thymocytes, peripheral T cytotoxic/suppressor cells, and most T helper cells, but not resting conventional peripheral B cells. CD43 expression has also been detected on pluripotent hematopoietic stem cells and myeloid, lymphoid, and NK-cell progenitors in the bone marrow. Studies of CD43-deficient mice indicate that CD43 participates in the negative regulation of T-cell activation and adhesion.
Spezifikation
CD43 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Mouse Plasmacytoma MOPC-315 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
S7 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified | |
Spn ; Sialophorin; Leukosialin; Ly-48; Ly48; Galgp; LEUK |
For Research Use Only.