Learn More
CD41a Mouse anti-Human, APC, Clone: HIP8, BD
Mouse Monoclonal Antibody
313.00 CHF - 650.00 CHF
Spezifikation
Antigen | CD41a |
---|---|
Klon | HIP8 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | APC |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15871399
|
BD Biosciences
561852 |
25 Tests |
313.00 CHF
1 Stück |
Verfügbar ab: 23-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15819778
|
BD Biosciences
559777 |
100 Tests |
650.00 CHF
1 Stück |
Verfügbar ab: 23-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The HIP8 monoclonal antibody specifically binds to the α-chain of CD41. CD41 is also known as Integrin αIIb or Platelet GPIIb. The calcium-dependent complex of CD41 and CD61 (β3 integrin or GPIIIa) is normally expressed on platelets and megakaryocytes. The CD41/CD61 complex is the receptor for fibrinogen, fibronectin and von Willebrand factor, and mediates platelet adhesion and aggregation. CD41 (clone HIP8) completely inhibits ADP-, epinephrine-, and collagen-induced platelet activation, and partially inhibits ristocetin- and thrombin-induced platelet activation. This antibody is useful in the morphological and physiological studies of platelets and megakaryocytes.
Spezifikation
CD41a | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
HIP8 | |
Monoclonal | |
Mouse | |
Human | |
Purified platelet membrane glycoproteins | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.