missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40 / TNFRSF5 Mouse, Clone: C40/1605, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 958-MSM1-P0
Additional Details : Gewicht : 0.01000kg
Spezifikation
CD40 / TNFRSF5 | |
Cell Surface and Secreted | |
C40/1605 | |
Unconjugated | |
XP_016883624 | |
Mouse | |
43kDa | |
Protein A/G Purified | |
RUO | |
958 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CD40 | |
Recombinant human full-length CD40 protein | |
L-428 Cells. Tonsil, Spleen or Thymus | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |