missing translation for 'onlineSavingsMsg'
Learn More

CD4 Rat anti-Mouse, Alexa Fluor™ 647, Clone: RM4-5, Invitrogen™

Rat Monoclonal Antibody

Marke:  Invitrogen MCD0421

Additional Details : Gewicht : 0.50000kg

Artikelnummer. 10232063

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen »


Beschreibung

Beschreibung

The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].
Spezifikation

Spezifikation

CD4
Monoclonal
0.1 mg/mL
PBS with 4mg/mL BSA and 0.1% sodium azide
P06332
Cd4
Mouse CD4
1 mL
Primary
Mouse
Antibody
IgG2a
Flow Cytometry
RM4-5
Alexa Fluor 647
Cd4
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rat
Purified
RUO
12504
4° C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote