missing translation for 'onlineSavingsMsg'
Learn More

CD4 Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 19068-1-AP-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16828854

  • 381.34 CHF / 150 Mikroliter
Verfügbar ab: 15-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The CD4 gene encodes a membrane glycoprotein of T lymphocytes that interacts with major histocompatibility complex class II antigens and is also a receptor for the human immunodeficiency virus. This gene is expressed not only in T lymphocytes, but also in B cells, macrophages, and granulocytes. It is also expressed in specific regions of the brain. The protein functions to initiate or augment the early phase of T-cell activation, and may function as an important mediator of indirect neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcript variants encoding different isoforms have been identified in this gene.
Spezifikation

Spezifikation

CD4
Polyclonal
Unconjugated
CD4
CD4, CD4 molecule, CD4mut
Rabbit
Antigen Affinity Chromatography
RUO
12504, 24932, 404704, 920
-20°C
Liquid
Immunoprecipitation, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P01730, P05540, P06332
CD4
CD4 Fusion Protein Ag13616
150 μL
Primary
Human, Mouse, Porcine, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote