missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19068-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The CD4 gene encodes a membrane glycoprotein of T lymphocytes that interacts with major histocompatibility complex class II antigens and is also a receptor for the human immunodeficiency virus. This gene is expressed not only in T lymphocytes, but also in B cells, macrophages, and granulocytes. It is also expressed in specific regions of the brain. The protein functions to initiate or augment the early phase of T-cell activation, and may function as an important mediator of indirect neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcript variants encoding different isoforms have been identified in this gene.Spezifikation
CD4 | |
Polyclonal | |
Unconjugated | |
CD4 | |
CD4, CD4 molecule, CD4mut | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12504, 24932, 404704, 920 | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P01730, P05540, P06332 | |
CD4 | |
CD4 Fusion Protein Ag13616 | |
20 μL | |
Primary | |
Rat, Human, Porcine, Mouse | |
Liquid |