missing translation for 'onlineSavingsMsg'
Learn More

CD4 Monoclonal Antibody (S3.5), Alexa Fluor™ 700, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MHCD0429

Additional Details : Gewicht : 0.50000kg

Artikelnummer. 10281723

  • 738.00 CHF / 1 Stück
Verfügbar ab: 27-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The Alexa Fluor® 700 dye conjugate provides an excellent spectral match to the common long-wavelength excitation sources, with a high extinction coefficient.

CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry.
Spezifikation

Spezifikation

CD4
Monoclonal
Alexa Fluor 700
CD4
Activation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen
Mouse
purified
ASR
920
4° C, store in dark
Liquid
Flow Cytometry
S3.5
PBS with 4mg/mL BSA and 0.1% sodium azide
P01730
CD4
Human CD4
500 μL
Primary
Human
Antibody
IgG2a
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote