missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3e Armenian Hamster, Biotin, Clone: 145-2C11, enQuireBio™
Armenian Hamster Monoclonal Antibody
Marke: EnquireBio QAB2-B-25ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
CD3e | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Cd3e | |
Protein A/G Purified | |
RUO | |
12501 | |
IgG |
Flow Cytometry | |
145-2C11 | |
Biotin | |
XP_006510029 | |
Armenian Hamster | |
25 μg | |
Primary | |
Store at 2-4°C. |