missing translation for 'onlineSavingsMsg'
Learn More

CD3e Armenian Hamster anti-Mouse, Super Bright 600, Clone: 145-2C11, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody

Marke:  Invitrogen 63-0031-82

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983454

  • 322.00 CHF / 100 Mikrogramm


Verfügbar ab
08-10-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

CD3e Monoclonal antibody specifically detects CD3e in Mouse samples. It is validated for Flow Cytometry
Spezifikation

Spezifikation

CD3e
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
P04235, P11942, P22646, P24161
4930549J05Rik; A430104F18Rik; AI504783; AW552088; BOS_14337; BOS_2540; Cd247; CD3; Cd3d; CD3-DELTA; Cd3e; CD3epsilon; Cd3-eta; Cd3g; CD3-GAMMA; Cd3h; CD3Q; CD3Z; Cd3zeta; CD3-ZETA; Ctg3; Ctg-3; FLJ18683; IMD17; IMD18; IMD19; IMD25; Leu-4; T3/TCR complex; T3D; T3e; T3G; T3Z; T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; T-cell receptor T3 zeta chain; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 delta chain; T-cell surface glycoprotein CD3 epsilon chain; T-cell surface glycoprotein CD3 gamma chain; T-cell surface glycoprotein CD3 zeta chain; TCRE; Tcrk; Tcrz; TCRzeta
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
145-2C11
Super Bright 600
Cd3d
Liquid
Cd247, Cd3d, CD3E, CD3g
IgG
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12500, 12501, 12502, 12503
Videos
Dokumentation