Learn More
CD38 Mouse anti-Human, FITC, Clone: HIT2, BD
Mouse Monoclonal Antibody
208.00 CHF - 366.00 CHF
Spezifikation
Antigen | CD38 |
---|---|
Klon | HIT2 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | FITC |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15830659
|
BD Biosciences
560982 |
25 Tests |
208.00 CHF
1 Stück |
Verfügbar ab: 23-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15838188
|
BD Biosciences
555459 |
100 Tests |
366.00 CHF
1 Stück |
Verfügbar ab: 23-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The HIT2 monoclonal antibody specifically binds to CD38. The CD38 antigen is also known as T10, ADP-ribosyl cyclase 1, and cyclic ADP ribose hydrolase 1. CD38 is a 45 kDa type II single-chain transmembrane glycoprotein present on thymocytes, activated T cells and terminally differentiated B cells (plasma cells). CD38 is expressed by other cells including monocytes, macrophages, dendritic cells, NK cells, myeloid and erythroid precursors and some epithelial cells. The CD38 antigen acts as an ectoenzyme that catalyzes the synthesis and hydrolysis of a Ca++ mobilizing agent, cyclic ADP-ribose. This intracellular calcium plays an important role in cell signaling pathways leading to cellular growth, apoptosis, and differentiation. CD38 binds to CD31 and thus plays a role in lymphocyte adhesion to endothelial cells.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Spezifikation
CD38 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
HIT2 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.