missing translation for 'onlineSavingsMsg'
Learn More

CD38 Mouse anti-Human, BUV496, Clone: HIT2, BD Horizon™

Mouse Monoclonal Antibody

Marke:  BD Biosciences 564657

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15833789

  • 650.00 CHF / 1 Stück


Verfügbar ab
28-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow cytometry.

CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow cytometry.
Spezifikation

Spezifikation

CD38
HIT2
Vol. Per test: 5μL
Purified
Murine
100 Tests
Cell Biology
Primary
Human
Flow Cytometry
Brilliant Ultraviolet 496
Aqueous buffered solution containing ≤0.09% sodium azide.
T10; ADP-ribosyl cyclase 1; Cyclic ADP-ribose hydrolase 1; gp45
Mouse IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimum conditions, and unconjugated antibody and free BD Horizon BUV496 were removed.
Monoclonal
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

For Research Use Only.