missing translation for 'onlineSavingsMsg'
Learn More

CD36 Mouse anti-Human, Clone: 1A8C5, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 66395-1-IG-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838235

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

CD36 (fatty acid translocase, FAT) is an 88 kDa, ditopic glycosylated protein that belongs to the class B family of scavenger receptors. CD36 is expressed by most resting marginal zone B cells but not by follicular and B1 B cells, and is rapidly induced on Follicular B cells in vitro upon TLR and CD40 stimulation. CD36 does not affect the development of B cells, but modulates both primary and secondary antibody response. Similar to glucose transporter GLUT4, CD36 is translocated from intracellular pools to the plasma membrane following cell stimulation by insulin. In mouse, CD36 is responsible for gustatory perception of long-chain fatty acids. CD36 is preferentially found within lipid rafts, which facilitates its association with receptors, signaling and adapter molecules. CD36 binds to collagen, thrombospondin, anionic phospholipids and oxidized LDL. Further, CD36 may function as a cell adhesion molecule and directly mediates cyto-adherence of Plasmodium falciparum parasitized erythrocytes. Mutations in the CD36 gene cause platelet glycoprotein deficiency. Multiple alternatively spliced transcript variants encoding the CD36 protein have been found.
Spezifikation

Spezifikation

CD36
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P16671
CD36
CD36 Fusion Protein Ag13541
20 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
1A8C5
Unconjugated
CD36
CD36, CHDS7, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, PAS 4, PAS IV, PASIV, Platelet collagen receptor, Platelet glycoprotein 4, Platelet glycoprotein IV, SCARB3, Thrombospondin receptor
Mouse
Protein G
RUO
948
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote