missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Antibody (D-2712) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 8 publications
265.00 CHF - 622.00 CHF
Spezifikation
| Antigen | CD36/SR-B3 |
|---|---|
| Klon | D-2712 |
| Konzentration | 1.0 mg/mL |
| Verdünnung | Western Blot 1:500, Immunohistochemistry 1:100-1:250, Immunoprecipitation 10-20 ug, Immunohistochemistry-Paraffin 1:100-1:250 |
| Anwendungen | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18706983
|
Novus Biologicals
NB110-59724SS |
0.025 mL |
265.00 CHF
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18707123
|
Novus Biologicals
NB110-59724 |
0.1 mL |
622.00 CHF
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
CD36 Monoclonal specifically detects CD36 in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Spezifikation
| CD36/SR-B3 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
| CD36 | |
| IgA κ | |
| Protein G purified | |
| Mouse and rat. Does not react with human. |
| D-2712 | |
| Western Blot 1:500, Immunohistochemistry 1:100-1:250, Immunoprecipitation 10-20 ug, Immunohistochemistry-Paraffin 1:100-1:250 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat, Human (Negative) | |
| 948 | |
| Adenovirus expressing recombinant mouse CD36. [Uniprot: Q08857] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 75 kDa |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts