missing translation for 'onlineSavingsMsg'
Learn More

CD357 (AITR/GITR) Rat anti-Mouse, Super Bright 702, Clone: DTA-1, eBioscience™

Rat Monoclonal Antibody

Marke:  Invitrogen 67-5874-80

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983764

  • 171.00 CHF / 25 Mikrogramm


Verfügbar ab
30-09-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product No. SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 702 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. Glucocorticoid-induced tumor necrosis factor receptor family-related gene (GITR) is a member of the tumor necrosis factor receptor (TNFR) family that is expressed at low levels on unstimulated T cells, B cells, and macrophages. Upon activation, CD4+ and CD8+ T cells up-regulate GITR expression, whereas immunoregulatory T cells constitutively express high levels of GITR.
Spezifikation

Spezifikation

CD357 (AITR/GITR)
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
O35714
AITR; CD357; GITR-D; Glucocorticoid-induced TNFR-related protein; TNFRSF18; Tumor necrosis factor receptor superfamily member 18
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
DTA-1
Super Bright 702
Tnfrsf18
Liquid
Tnfrsf18
IgG2b, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
21936
Videos
Dokumentation