missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Antibody (1H6) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals FAB3346B
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD34 Monoclonal antibody specifically detects CD34 in Canine samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Spezifikation
| CD34 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Canine | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 1H6 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
| COS-7 African green monkey SV40 transformed kidney fibroblast-like cell line-derived recombinant canine CD34 | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
| 947 | |
| Store at 4°C in the dark. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur