missing translation for 'onlineSavingsMsg'
Learn More

CD335 (NKp46) Rat anti-Mouse, Super Bright 645, Clone: 29A1.4, eBioscience™

Rat Monoclonal Antibody

Marke:  Invitrogen 64-3351-80

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983644

  • 161.00 CHF / 25 Mikrogramm


Verfügbar ab
07-10-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product No. SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 645 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. Cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis.
Spezifikation

Spezifikation

CD335 (NKp46)
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
Q149L7
CD355; class I MHC restricted T cell associated molecule; class-I MHC-restricted T cell associated molecule; class-I MHC-restricted T-cell-associated molecule; cytotoxic and regulatory T cell molecule; NKp46
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
29A1.4
Super Bright 645
Crtam
Liquid
Crtam
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
54698
Videos
Dokumentation