missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD335 (NKp46) Monoclonal Antibody (VIV-KM1), APC, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA528352
Additional Details : Gewicht : 0.01000kg
Beschreibung
This antibody recognizes ovine and caprine CD335. Reconstitute with 1.0 mL distilled water.
NKp46 (CD335, NCR1, ) is a cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis. NKp46 consists of two Ig-like domains assembled to leukocyte immunoglobulin-like (LIR) and killer inhibitory receptors (KIR).Spezifikation
CD335 (NKp46) | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
0 | |
NCR1 | |
Fusion protein consisting of the extracellular region of porcine CD335 | |
100 Tests | |
Primary | |
Porcine | |
Antibody | |
IgG1 |
Flow Cytometry | |
VIV-KM1 | |
APC | |
NCR1 | |
1-Mar; activating receptor 1; Ar1; CD335; hNKp46; Kilr1; KILR-1; LY94; Ly-94; lymphocyte antigen 94; Lymphocyte antigen 94 homolog; lymphocyte antigen 94 homolog (activating NK-receptor; lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46); mAR-1; MAR1 (mouse activating receptor 1); mNKp46; natural cytotoxicity triggering receptor 1; Natural killer cell p46-related protein; natural killer cell receptor; Ncr1; NK cell-activating receptor; NK receptor KILR-1; NKACTR; Nkp46; NK-p46; NK-p46); rAR-1; rat-activating receptor 1 | |
Mouse | |
Protein A | |
RUO | |
100141419 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |