missing translation for 'onlineSavingsMsg'
Learn More

CD324 (E-Cadherin) Monoclonal Antibody (DECMA-1), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

216.00 CHF - 466.00 CHF

Spezifikation

Antigen CD324 (E-Cadherin)
Klon DECMA-1
Konzentration 0.5 mg/mL
Anwendungen Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen)
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15322020
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
53-3249-80
25 μg
216.00 CHF
25 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15332020
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
53-3249-82
100 μg
466.00 CHF
100 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

Description: The monoclonal antibody DECMA-1 recognizes mouse, human and canine CD324 also known as E-cadherin (Epithelial cadherin) or uvomorulin. Like the other cadherin family members P and N cadherin, E-cadherin is a transmembrane glycoprotein involved in intercellular adhesion. These proteins share a common basic structure. The extracellular portions of the proteins are largely composed of repeating domains, each with two consensus Ca2+-binding motifs. The cytoplasmic domain interacts with a-, b-, and g-catenins and actinins. These catenins connect E-cadherin with the cytoskeleton. Expression is found in most epidermal cells including melanocytes and kerotinocytes. E-cadherin is localized at the intercellular boundaries of epithelial cells in several tissues, and is thought to play a role in maintece of tissue integrity. Loss of E-cadherin function has been implicated in the progression of a variety of cancers. E-Cadherin protein is sensitive to trypsin treatment, so exposure to trypsin should be minimized or avoided. The monoclonal antibody DECMA-1 has been shown to have functional activity by disrupting adhesion in human, mouse and dog cells. Applications Reported: This DECMA-1 antibody has been reported for use in flow cytometric analysis, immunohistochemical staining of frozen tissue sections, and immunocytochemistry.

CD324 (E-Cadherin) Monoclonal antibody specifically detects CD324 (E-Cadherin) in Canine, Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen).
Spezifikation

Spezifikation

CD324 (E-Cadherin)
0.5 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
AA960649; ARC-1; cadherin 1; cadherin 1, E-cadherin (epithelial); cadherin 1, epithelial; cadherin 1, type 1; cadherin 1, type 1, E-cadherin (epithelial); cadherin e; cadherin-1; cadherin-E; calcium-dependent adhesion protein, epithelial; CAM 120/80; CD324; CDH1; CDHE; cell adhesion molecule; cell-CAM 120/80; ECAD; E-cad; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3; Ecadherin; E-cadherin; E-cadherin 1; E-cadherin precursor; Epithelial cadherin; hab; half baked; LCAM; L-CAM; Um; UVO; Uvomorulin
CDH1
Primary
4° C, store in dark, DO NOT FREEZE!
CDH1
DECMA-1
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen)
Alexa Fluor 488
Rat
Canine, Human, Mouse
F1PAA9, P09803, P12830
12550, 442858, 999
IgG1 κ
Affinity chromatography
Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications

For Research Use Only.