missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD32 Rat, Clone: 2.4G2, enQuireBio™
Rat Monoclonal Antibody
Marke: EnquireBio QAB87-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
CD32 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
FCGR2b | |
Protein A/G Purified | |
RUO | |
14131 | |
IgG2b |
Flow Cytometry | |
2.4G2 | |
Unconjugated | |
XP_006496722 | |
Rat | |
1 mg | |
Primary | |
Store at 2-4°C. |