Learn More
CD283 (TLR3) Monoclonal Antibody (TLR3.7), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
256.00 CHF - 452.00 CHF
Spezifikation
Antigen | CD283 (TLR3) |
---|---|
Klon | TLR3.7 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15229349
|
Affymetrix eBioscience
12-9039-80 |
25 μg |
256.00 CHF
25 Mikrogramm |
Verfügbar ab: 02-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15239349
|
Affymetrix eBioscience
12-9039-82 |
100 μg |
452.00 CHF
100 Mikrogramm |
Verfügbar ab: 02-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Description: The TLR3.7 monoclonal antibody reacts with human Toll-like receptor 3 (TLR3). To date, at least twelve members of the Toll-like receptor family have been identified. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. In the innate immune response, TLRs recognize molecular patterns associated with microbial pathogens and induce antimicrobial activity. TLR3 recognizes double-stranded (ds)RNA, induces the activation of NF-kappaB through MyD88-dependent and -independent pathways, and the production of type I interferons (IFNs). TLR3.7 suppressed poly(I):poly(C)-mediated IFN-beta production by human fibroblasts naturally expressing TLR3 on their surface. Applications Reported: The TLR3.7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TLR3.7 antibody has been tested by intracellular staining and flow cytometric analysis of A549 and MRC5 cell lines to detect the very low level of this antigen on the surface. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
TLR3 (Toll-like receptor 3, CD283) is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from drosophila to humans and share structural and functional similarities. TLRs recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. TLR3 is most abundantly expressed in placenta and pancreas, and is restricted to the dendritic subpopulation of the leukocytes. TLR3 recognizes dsRNA associated with viral infection, and induces the activation of NF-kappaB and the production of type I interferons. Further, TLR3 may play a role in host defense against viruses, and the use of alternative polyadenylation sites to generate different length transcripts has been noted for the TLR3 gene. Like its counterparts in Drosophila, TLRs signal through adaptor molecules and could constitute an important and unrecognized component of innate immunity in humans. The TLR family is a phylogenetically conserved mediator of innate immunity that is essential for microbial recognition. TLRs characterized so far activate the MyD88/interleukin-1 receptor-associated kinase (IRAK) signaling pathway. TLR3 is the only human TLR which does not utilize MyD88 as an adaptor molecule instead utilizing TRIF in a signaling path which results in downstream activation of IRF-3 and NF-kB. TLR3 activation leads not only to Type I interferon induction but also to other inflammatory cytokines which result in DC maturation. TLR3 is known to recognize viral double-stranded (ds) RNA, a molecular pattern associated with viral infection. Recently it has been shown to recognize viruses such as Influenza A and West Nile Virus and can mediate entry of at least West Nile Virus.Spezifikation
CD283 (TLR3) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AI957183; CD283; IIAE2; TLR 3; TLR3; TLR-3; toll like receptor 3; toll-like receptor 3; toll-like receptor 3 variant 1 | |
TLR3 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
TLR3 |
TLR3.7 | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
O15455 | |
7098 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.