missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Mouse, APC, Clone: CD28.2, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio QAB37-APC-100Tests
Additional Details : Gewicht : 0.01000kg
Spezifikation
CD28 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
CD28 | |
Protein A/G Purified | |
RUO | |
940 | |
IgG1 κ |
Flow Cytometry | |
CD28.2 | |
APC | |
NP_006130 | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-4°C. |