Learn More
CD279 (PD-1) Rat anti-Mouse, PE, Clone: RMP1-30, Proteintech
Rat Monoclonal Antibody
Marke:  Proteintech PE-65142-100UG
Artikelnummer. 16801736
  • 101.79 CHF / 100 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
The CD279 (PD-1) antibody from Proteintech is a rat monoclonal antibody generated with mouse pd-1 transfected bhk cells of mouse CD279 (PD-1). This antibody recognizes mouse antigen. The CD279 (PD-1) antibody has been validated for the following applications: FC analysis. This antibody is pe.
Spezifikation
Spezifikation
CD279 (PD-1)
Monoclonal
0.2 mg/mL
Rat
Primary
Mouse
Flow Cytometry
RMP1-30
PE
100 μg
18566
IgG2b
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen