missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD279 (PD-1) Monoclonal Antibody (RMP1-14), eBioscience™, Invitrogen™

Rat Monoclonal Antibody

427.00 CHF

Spezifikation

Antigen CD279 (PD-1)
Klon RMP1-14
Konzentration 0.5 mg/mL
Anwendungen Flow Cytometry, Functional Assay
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 1
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15278307
Unterlagen ansehen
Invitrogen™
14-9982-85
500 μg
427.00 CHF
500 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung

Beschreibung

This antibody reacts with mouse PD-1 (programmed death-1), a 55 kDa member of the Ig superfamily. PD-1 contains the immunoreceptor tyrosine-based inhibitory motif (ITIM) and plays a key role in peripheral tolerance and autoimmune disease in mice. PD-1 is expressed mainly on activated T and B lymphocytes. Two novel B7 Family members have been identified as PD-1 ligands, PD-L1 (B7-H1) and PD-L2 (B7-DC). Evidence reported to date suggests overlapping functions for these ligands and their constitutive expression on some normal tissues and upregulation on activated antigen-presenting cells. Similar to J43, another mAb to mouse PD-1 (cat. 16-9985), RMP1-14 blocks the binding of both B7-H1-Ig and B7-DC-Ig to PD-1 transfectants.

Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of "e;danger signals"e; such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases. Despite its predicted molecular weight, PD-1 often migrates at higher molecular weight in SDS-PAGE.
Spezifikation

Spezifikation

CD279 (PD-1)
0.5 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2
Pdcd1
Primary
4°C
Pdcd1
RMP1-14
Flow Cytometry, Functional Assay
Unconjugated
Rat
Mouse
Q02242
18566
IgG2a κ
Affinity chromatography
Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt