missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD279 (PD-1) Monoclonal Antibody (J43), Super Bright™ 645, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Marke: Invitrogen 64-9985-80
Additional Details : Gewicht : 0.01000kg
Beschreibung
Armenian Hamster Monoclonal Antibody
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Mouse samples. It is validated for Flow CytometrySpezifikation
CD279 (PD-1) | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Q02242 | |
Pdcd1 | |
Affinity chromatography | |
RUO | |
18566 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
J43 | |
Super Bright 645 | |
Pdcd1 | |
CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; Pd1; PD-1; Pdc1; Pdcd1; Programmed cell death protein 1; Protein PD1; Protein PD-1; sCD279; SLEB2; soluble CD279 | |
Armenian Hamster | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |