Learn More
CD279 Mouse anti-Human, PE, Clone: MIH4, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 557946
Additional Details : Gewicht : 0.02000kg
Beschreibung
The MIH4 monoclonal antibody specifically binds to the program death 1 (PD-1) receptor CD279, a member of the Ig superfamily. CD279 is an immunoregulatory receptor expressed on activated T cells, B cells and myeloid cells. Mice deficient in CD279 show a breakdown of peripheral tolerance and manifest multiple autoimmune symptoms. It contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) in the cytoplasmic region. PD-L1 and PD-L2 are ligands of CD279 and are members of the B7 gene family. Interaction of CD279:PD-Ligands results in inhibition of T cell proliferation and cytokine secretion. Reports suggest that the B7/CTLA-4 pathway functions primarily to attenuate, limit, and/or terminate naοve T-cell activation in secondary lymphoid organs. The PD-ligand:CD279 pathway, on the other hand, may primarily attenuate, limit, and/or terminate T-, B-, and myeloid cell activation/effector function at sites of inflammation in the periphery.
Spezifikation
CD279 | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human PD-1 Transfected Cell Line | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
MIH4 | |
PD-1; hPD-1; PD1; PDCD1; Programmed cell death 1; SLEB2 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.