Learn More
CD279 Mouse anti-Human, FITC, Clone: MIH4, BD
Mouse Monoclonal Antibody
204.00 CHF - 415.00 CHF
Spezifikation
Antigen | CD279 |
---|---|
Klon | MIH4 |
Konzentration | 0.5mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15830709
|
BD Biosciences
561035 |
25 μg |
204.00 CHF
1 Stück |
Verfügbar ab: 22-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15879078
|
BD Biosciences
557860 |
0.1 mg |
415.00 CHF
1 Stück |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The MIH4 monoclonal antibody specifically binds to the program death 1 (PD-1) receptor CD279, a member of the Ig superfamily. CD279 is an immunoregulatory receptor expressed on activated T cells, B cells and myeloid cells. Mice deficient in CD279 show a breakdown of peripheral tolerance and manifest multiple autoimmune symptoms. It contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) in the cytoplasmic region. PD-L1 and PD-L2 are ligands of CD279 and are members of the B7 gene family. Interaction of CD279:PD-Ligands results in inhibition of T cell proliferation and cytokine secretion. Reports suggest that the B7/CTLA-4 pathway functions primarily to attenuate, limit, and/or terminate naοve T-cell activation in secondary lymphoid organs. The PD-ligand:CD279 pathway, on the other hand, may primarily attenuate, limit, and/or terminate T-, B-, and myeloid cell activation/effector function at sites of inflammation in the periphery.
Spezifikation
CD279 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Human | |
Human PD-1 Transfected Cell Line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
MIH4 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
PD-1; hPD-1; PD1; PDCD1; Programmed cell death 1; SLEB2 |
For Research Use Only.