missing translation for 'onlineSavingsMsg'
Learn More

CD278 (ICOS), Super Bright 436, clone: ISA-3, eBioscience™

Mouse Monoclonal Antibody

Marke:  Invitrogen 62-9948-41

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15815543

  • 158.00 CHF / 25 Stück


Geschätzter Ausliefertermin
von Anbieter 06-05-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Applications Tested: This ISA-3 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (cat. SB-4400) to minimize any non-specific polymer interactions. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser CD278 belongs to the CD28 and CTLA-4 cell-surface receptor family and is highly expressed in activated T-cells. It is essential both for efficient interaction between T and B-cells and for normal antibody responses to T-cell dependent antigens. It enhances all basic T-cell responses to a foreign antigen, namely proliferation, secretion of lymphokines, up-regulation of molecules that mediate cell-cell interaction, and effective help for antibody secretion by B-cells.
Spezifikation

Spezifikation

CD278 (ICOS)
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
Q9Y6W8
ICOS
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
29851
Flow Cytometry
ISA-3
Super Bright 436
ICOS
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos

For Research Use Only