missing translation for 'onlineSavingsMsg'
Learn More

CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH5), Functional Grade, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

161.00 CHF - 470.00 CHF

Spezifikation

Antigen CD274 (PD-L1, B7-H1)
Klon MIH5
Konzentration 1 mg/mL
Anwendungen Flow Cytometry, Functional Assay, Neutralization
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 3
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15278717
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
16-5982-81
50 μg
161.00 CHF
50 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 07-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15288717
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
16-5982-82
100 μg
313.00 CHF
100 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15298717
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
16-5982-85
500 μg
470.00 CHF
500 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

Description: The MIH5 monoclonal antibody reacts with mouse B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40 kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes including T, B, NK and DC. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. MIH5 is reported to be a blocking antibody. Applications Reported: The MIH5 antibody has been reported for use in flow cytometric analysis. It has also been reported in blocking in in vitro and in vivo functional assays. Applications Tested: The MIH5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.
Spezifikation

Spezifikation

CD274 (PD-L1, B7-H1)
1 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.2
A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1
Cd274
Primary
4° C
Cd274
MIH5
Flow Cytometry, Functional Assay, Neutralization
Functional Grade
Rat
Mouse
Q9EP73
60533
IgG2a λ
Affinity chromatography
Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications

For Research Use Only.