Learn More
CD25 Monoclonal Antibody (PC61.5), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
170.00 CHF - 256.00 CHF
Spezifikation
Antigen | CD25 |
---|---|
Klon | PC61.5 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
12730110
|
Affymetrix eBioscience
25-0251-81 |
50 μg |
170.00 CHF
1 Stück |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
15330910
|
Affymetrix eBioscience
25-0251-82 |
100 μg |
256.00 CHF
100 Mikrogramm |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
Beschreibung
Description: The PC61.5 antibody reacts with mouse CD25, the 55 kDa interleukin-2 receptor alpha chain (IL-2R alpha). CD25 is expressed by early progenitors of the T and B lineage as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 only with low affinity. However, CD25 associates with CD122 (IL-2 receptor beta chain) and CD132 (common gamma chain) to form the high affinity IL-2 receptor. Binding of IL-2 to both the high and low affinity classes of IL-2 receptor is inhibited by the PC61.5 antibody. CD25 plays a role in lymphocyte differentiation and activation/proliferation. Applications Reported: The PC61.5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This PC61.5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD25 Monoclonal antibody specifically detects CD25 in Mouse samples. It is validated for Flow Cytometry.Spezifikation
CD25 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
ADA; ADA1; Adenosine aminohydrolase; adenosine deaminase; CD25; CD25 antigen; cytokine receptor; IDDM10; IL 2 RA; IL 2 receptor; IL 2R; IL 2R subunit alpha; IL2 RA; IL2 RA antibody; IL2 receptor; il-2 receptor alpha; IL-2 Receptor alpha chain; IL-2 receptor alpha subunit; IL2 receptor subunit alpha; IL-2 receptor subunit alpha; IL2R; IL-2R alpha; IL-2R alpha chain; IL2R subunit alpha; IL-2R subunit alpha; Il2ra; IL-2RA; IL-2-RA; IL2-RA; Il2ra antibody; IL2RAC; IMD41; interleukin 2 receptor; interleukin 2 receptor alpha chain; interleukin 2 receptor subunit alpha; interleukin 2 receptor, alpha; interleukin 2 receptor, alpha chain; interleukin-2 receptor alpha; interleukin-2 receptor alpha chain; interleukin-2 receptor alpha subunit; interleukin-2 receptor beta chain; Interleukin2 receptor subunit alpha; interleukin-2 receptor subunit alpha; Ly-43; p55; p55 chain; sIL 2R; soluble IL 2 receptor; TAC antigen; TCGFR | |
IL2RA | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IL2RA |
PC61.5 | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Mouse | |
P01590 | |
16184 | |
IgG1 λ | |
Affinity chromatography | |
Antibody |
For Research Use Only.