Learn More
CD25 Rat anti-Mouse, PE, Clone: PC61, BD
Rat Monoclonal Antibody
154.00 CHF - 515.00 CHF
Spezifikation
Antigen | CD25 |
---|---|
Klon | PC61 |
Konzentration | 0.2mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15830739
|
BD Biosciences
561065 |
25 μg |
154.00 CHF
1 Stück |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15847438
|
BD Biosciences
553866 |
0.2 mg |
515.00 CHF
1 Stück |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The PC61 antibody reacts with CD25, the low-affinity IL-2 Receptor α chain (IL-2Rα, p55) expressed on activated T and B lymphocytes from all mouse strains tested. IL-2Rα by itself is not a signaling receptor. However, it can combine with IL-2 Receptor β (CD122) and γc (CD132) chains to form high-affinity, signaling receptor complexes for IL-2. Resting T and B lymphocytes and resting and activated NK cells do not express IL-2Rα. CD25 is transiently expressed at a low level during normal B-cell development in the bone marrow on the CD45R/B220low TdT- sIg- Pre-B/Pre-B-II and CD45R/B220low TdT- sIgM+ sIgD- immature B stages, but not on the CD45R/B220low TdT+ sIg- Pro-B/Pre-B-I stage nor on CD45R/B220high TdT- sIgM+ sIgD+ mature B cells. It is expressed at a higher level during a very early stage of T-cell development in fetal and adult thymus. Peripheral CD25+CD4+ lymphocytes called regulatory T (Treg) cells are involved in the maintenance of self-tolerance. It has also been reported that dendritic cells express CD25, recognized by mAb 7D4 (Cat. No. 553068). The PC61 antibody recognizes an epitope of CD25 which is distinct from the IL-2 binding site and from those recognized by mAbs 3C7 (Cat. No. 557364) and 7D4 (Cat. No. 553068). It blocks binding of IL-2 to CD25, presumably by inducing a conformational change in CD25.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Spezifikation
CD25 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
IL-2-dependent cytolytic mouse T-cell clone B6.1 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
PC61 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 λ | |
Affinity Purified | |
Interleukin-2 receptor alpha chain; IL-2RA; IL-2Rα; Il2ra; IL-2R p55 |
For Research Use Only.