missing translation for 'onlineSavingsMsg'
Learn More

CD244.2 (2B4) Rat anti-Mouse, FITC, Clone: eBio244F4, eBioscience™

Rat Monoclonal Antibody

Marke:  Affymetrix eBioscience 11-2441-81

Additional Details : Gewicht : 0.25000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15228199

  • 90.75 CHF / 50 Mikrogramm


Verfügbar ab
21-07-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Applications Tested: This eBio244F4 antibody has been tested by flow cytometric analysis of IL-2-stimulated mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. This gene encodes a cell surface receptor expressed on natural killer (NK) cells (and some T cells) that mediate non-major histocompatibility complex (MHC) restricted killing. The interaction between NK-cell and target cells via this receptor is thought to modulate NK-cell cytolytic activity. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Spezifikation

Spezifikation

CD244.2 (2B4)
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q07763
Cd244
IgG2a
50 μg
4° C, store in dark, DO NOT FREEZE!
Primary
18106
Flow Cytometry
eBio244F4
FITC
Cd244
Liquid
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Videos
Dokumentation

For Research Use Only.