Learn More
CD235a Monoclonal Antibody (YTH89.1), APC, Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA517697
Additional Details : Gewicht : 0.01000kg
Beschreibung
MA5-17697 targets CD235a/Glycophorin A in FACS and WB applications and shows reactivity with Human samples. The MA5-17697 immunogen is rat monoclonal antibodies raised against human CD235a.
CD235a (Glycophorins A, GPA) is a single pass membrane sialoglycoprotein expressed in mature erythrocytes and erythroid precursor cells. CD235a is the carrier of blood group M and N specificities. CD235a provides cells with a large mucin-like that may serve as a barrier to cell fusion, minimizing aggregation between red blood cells in the circulation. CD235a has been shown to act as a receptor for Sandei virus, parvovirus, and Hsa, an Streptococcus adhesin.Spezifikation
CD235a | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P02724 | |
GYPA | |
Rat monoclonal antibodies raised against human CD235a. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry | |
YTH89.1 | |
APC | |
GYPA | |
AI853584; CD235; CD235a; erythroid-lineage-specific membrane sialoglycoprotein; glycophorin A; glycophorin A (MN blood group); glycophorin A (MNS blood group); glycophorin A, GPA; glycophorin Erik; glycophorin MiI; glycophorin MiV; glycophorin SAT; glycophorin Sta type C; glycophorin-A; GPA; GPErik; GPSAT; Gypa; HGNC:4702; HGpMiV; HGpMiX; HGpMiXI; HGpSta(C); HGpStaC; Mi.V glycoprotein; Mi.V glycoprotein (24 AA); MN; MN sialoglycoprotein; MNS; PAS-2; recombinant glycophorin A-B Miltenberger-DR; sialoglycoprotein alpha | |
Rat | |
Protein G | |
RUO | |
2993 | |
4° C, do not freeze | |
Liquid |