missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD223 Mouse anti-Human, Alexa Fluor 647, Clone: T47-530, BD Pharmingen™
Mouse Monoclonal Antibody
Marke: BD Biosciences 565716
Additional Details : Gewicht : 0.00900kg
Beschreibung
CD223 Monoclonal antibody specifically detects CD223 in Human samples. It is validated for Flow cytometry.
CD223 Monoclonal antibody specifically detects CD223 in Human samples. It is validated for Flow cytometry.Spezifikation
CD223 | |
Monoclonal | |
Alexa Fluor 647 | |
LAG3; CD223; FDC; Lymphocyte activation gene 3 protein; Protein FDC | |
Human LAG-3 Recombinant Protein | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed. | |
IgG1 κ |
Flow Cytometry | |
T47-530 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Cell Biology | |
Human | |
Purified |
For Research Use Only.