missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD223 Mouse anti-Human, Alexa Fluor 647, Clone: T47-530, BD Pharmingen™
Mouse Monoclonal Antibody
273.00 CHF - 590.00 CHF
Spezifikation
Antigen | CD223 |
---|---|
Klon | T47-530 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | Alexa Fluor 647 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15844489
|
BD Biosciences
565717 |
25 Tests |
273.00 CHF
1 Stück |
Verfügbar ab: 22-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15834489
|
BD Biosciences
565716 |
100 Tests |
590.00 CHF
1 Stück |
Verfügbar ab: 22-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
CD223 Monoclonal antibody specifically detects CD223 in Human samples. It is validated for Flow cytometry.
CD223 Monoclonal antibody specifically detects CD223 in Human samples. It is validated for Flow cytometry.Spezifikation
CD223 | |
Flow Cytometry | |
Alexa Fluor 647 | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human LAG-3 Recombinant Protein | |
Primary |
T47-530 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
LAG3; CD223; FDC; Lymphocyte activation gene 3 protein; Protein FDC | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed. |
For Research Use Only.