Learn More
CD21/CD35 Monoclonal Antibody (eBio8D9 (8D9)), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marke: Affymetrix eBioscience 14-0211-95
Beschreibung
The MAb eBio8D9 reacts with an eptiope shared by mouse CD21 (CR2) and CD35 (CR1). CD21 and CD35 are alternatively spliced transcripts from the Cr2 gene, which produce cell-surface proteins of 145 and 190 kDa, respectively. CD21 and CD35 are expressed by mature B cells, but not on thymocytes, peripheral T cells, erythrocytes or platelets. Furthermore, there is some evidence which demonstrates their expression on macrophages. CD21 is a receptor for the complement component C3d and Epstein-Barr virus (EBV). In association with CD19 and CD81, CD21 also participates in B-cell activation through the B cell receptor. Cr2-deficient mice display impaired inflammatory and humoral immune responses in vivo. The anti-mouse CD21/35 MAb clones eBio4E3 and eBio8D9 do not cross-block each other, suggesting that they bind to different epitopes.
CD21/CD35 Monoclonal antibody specifically detects CD21/CD35 in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Western Blot
Spezifikation
| CD21/CD35 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P19070 | |
| Rat | |
| 10 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a λ |
| Flow Cytometry, Functional Assay, Western Blot | |
| eBio8D9 (8D9) | |
| Unconjugated | |
| Cr2 | |
| C3b/C4b receptor; C3-binding protein; C3DR; CD21; CD35; CD35 antigen; complement C3d receptor; complement receptor 2; complement receptor type 1; complement receptor type 2; Cr1; Cr-1; Cr2; Cr-2; EBV receptor; expressed in B lymphocytes | |
| Affinity chromatography | |
| RUO | |
| 12902 | |
| 4° C | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.