missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200R (soluble) Monoclonal Antibody (OX110), FITC, Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA517984
Additional Details : Gewicht : 0.01000kg
Beschreibung
MA5-17984 targets CD200R/OX-2R in FACS applications and shows reactivity with Mouse samples. The MA5-17984 immunogen is soluble recombinant mouse CD200R.
CD200R (OX2R) is a receptor for the OX-2 membrane glycoprotein, and both the receptor and substrate are cell surface glycoproteins containing two immunoglobulin-like domains. CD200R is restricted to the surfaces of myeloid lineage cells and the receptor-substrate interaction may function as a myeloid downregulatory signal. In mouse, CD200R is expressed primarily by peripheral blood monocytes and neutophils but also by other leucocytes including T-lymphocytes and mast cells. Further mouse studies of a gene related to CD200R suggests that the receptor may control myeloid function in a tissue-specific manner. Alternative splicing of CD200R results in multiple transcript variants. Diseases associated with CD200R dysfunction include primary hypertrophic osteoarthropathy.Spezifikation
CD200R (soluble) | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Q9ES57 | |
CD200R1 | |
Soluble recombinant mouse CD200R | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
OX110 | |
FITC | |
CD200R1 | |
antigen identified by monoclonal antibody MRC OX-2 receptor; CD200 cell surface glycoprotein receptor; CD200 R1; CD200 receptor 1; CD200R; Cd200r1; Cell surface glycoprotein CD200 receptor 1; cell surface glycoprotein OX2 receptor 1; cell surface glycoprotein receptor CD200; CRTR2; HCRTR2; MOX2 receptor; MOX2R; Orexin receptor 2; OX102 antigen; OX2 receptor; OX2R; UNQ2522/PRO6015 | |
Rat | |
Protein G | |
RUO | |
57781 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |