missing translation for 'onlineSavingsMsg'
Learn More

CD20 Mouse anti-Human Rhesus, Cynomolgus, Baboon, BUV805, Clone: 2H7, BD Horizon™

Mouse Monoclonal Antibody

Marke:  BD Biosciences 564918

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15833969

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.


Beschreibung

Beschreibung

CD20 Monoclonal antibody specifically detects CD20 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.

CD20 Monoclonal antibody specifically detects CD20 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.
Spezifikation

Spezifikation

CD20
Monoclonal
Brilliant Ultraviolet 805
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
25 Tests
Cell Biology
Baboon, Cynomolgus, Human, Rhesus Monkey
Purified
Flow Cytometry
2H7
Vol. Per test: 5μL
MS4A1; B1; Bp35; LEU-16; S7
Human 6.16c1.3 B cell line
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV805 under optimum conditions and unconjugated antibody and free BD Horizon BUV805 were removed.
IgG2b κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.