missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Mouse anti-Human Rhesus, Cynomolgus, Baboon, BUV805, Clone: 2H7, BD Horizon™
Mouse Monoclonal Antibody
Marke: BD Biosciences 564917
Additional Details : Gewicht : 0.01000kg
Beschreibung
CD20 Monoclonal antibody specifically detects CD20 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.
CD20 Monoclonal antibody specifically detects CD20 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.Spezifikation
CD20 | |
Monoclonal | |
Brilliant Ultraviolet 805 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Cell Biology | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
Purified |
Flow Cytometry | |
2H7 | |
Vol. Per test: 5μL | |
MS4A1; B1; Bp35; LEU-16; S7 | |
Human 6.16c1.3 B cell line | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV805 under optimum conditions and unconjugated antibody and free BD Horizon BUV805 were removed. | |
IgG2b κ |
For Research Use Only.