missing translation for 'onlineSavingsMsg'
Learn More

CD19 Monoclonal Antibody (SJ25C1), Super Bright™ 645, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

168.00 CHF - 389.00 CHF

Spezifikation

Antigen CD19
Klon SJ25C1
Konzentration 5 μL/Test
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15993604
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
64-0198-42
100 Tests
389.00 CHF
100 Stück
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 10-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15983604
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
64-0198-41
25 Tests Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
N/A Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Beschreibung

Beschreibung

Description: The HIB19 monoclonal antibody reacts with human CD19, a 95 kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, Leu13, MHC class II, and CD19 form a multimolecular complex that associates with BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells. The SJ25C1 antibody and the HIB19 monoclonal antibody recognize overlapping epitopes. Applications Reported: This HIB19 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HIB19 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (cat.

CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry.
Spezifikation

Spezifikation

CD19
5 μL/Test
Monoclonal
Liquid
RUO
P15391
IgG1 κ
Affinity chromatography
Antibody
SJ25C1
Flow Cytometry
Super Bright 645
Mouse
Human
930
Primary
4° C, store in dark, DO NOT FREEZE!
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications