missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Rat, V450, Clone: 1D3, enQuireBio™
Rat Monoclonal Antibody
Marke: EnquireBio QAB28-V450-25ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
CD19 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Cd19 | |
Protein A/G Purified | |
RUO | |
12478 | |
IgG2a κ |
Flow Cytometry | |
1D3 | |
Violet Fluor 450 (V450) | |
NP_033974 | |
Rat | |
25 μg | |
Primary | |
Store at 2-4°C. |