missing translation for 'onlineSavingsMsg'
Learn More

CD19 Mouse anti-Human, BUV496, Clone: SJ25C1, BD Horizon™

Mouse Monoclonal Antibody

Marke:  BD Biosciences 564656

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15823789

  • 307.00 CHF / 1 Stück


Verfügbar ab
01-06-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow cytometry.

CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow cytometry.
Spezifikation

Spezifikation

CD19
SJ25C1 (also known as SJ25-C1)
Vol. Per test: 5μL
Purified
Murine
Mouse BALB/c IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimum conditions, and unconjugated antibody and free BD Horizon BUV496 were removed.
Monoclonal
Flow Cytometry
Brilliant Ultraviolet 496
Aqueous buffered solution containing ≤0.09% sodium azide.
B4; B-lymphocyte antigen CD19; Leu-12
Human NALM1 + NALM16 cells
25 Tests
Cell Biology
Primary
Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

For Research Use Only.