Learn More
CD185 (CXCR5) Monoclonal Antibody (MU5UBEE), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00 CHF - 372.00 CHF
Spezifikation
Antigen | CD185 (CXCR5) |
---|---|
Klon | MU5UBEE |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15973444
|
Affymetrix eBioscience
62-9185-41 |
25 Tests |
178.00 CHF
25 Stück |
Verfügbar ab: 30-04-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15983444
|
Affymetrix eBioscience
62-9185-42 |
100 Tests |
372.00 CHF
100 Stück |
Verfügbar ab: 30-04-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Description: The MU5UBEE monoclonal antibody reacts with human and non-human primate CD185. CD185, which is also known as C-X-C chemokine receptor 5 (CXCR5) and Burkitt lymphoma receptor 1 (BLR1), is a seven transmembrane G protein-coupled receptor originally identified in Burkitt's lymphoma. In peripheral blood, CD185 is expressed on B cells, CD4+ T cells (but not Th1 or Th2 cells), as well as on a subpopulation of memory (CD45RO+) T cells. Circulating CD185+ T cells are in a resting state and migrate to the lymph nodes due to expression of CCR7 and CD62L. In tonsil, CD185 is expressed on nearly all CD4+ cells together with CD45RO and activation markers such as CD69 and ICOS. Tonsillar CD185+ cells have been shown to induce antibody production when co-cultured with B cells, thus supporting their role in providing help to B cells. Furthermore, this chemokine receptor plays a critical role in lymphocyte trafficking, in particular T cell migration into the B cell follicles of germinal centers in response to CXCL13, making CD185 an established marker of follicular helper T cells. Applications Reported: This MU5UBEE antibody has been reported for use in flow cytometric analysis. Applications Tested: This MU5UBEE antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (1.0 μg) per test.
CD185 (CXCR5) Monoclonal antibody specifically detects CD185 (CXCR5) in Human, Non-human Primate, Rhesus Monkey samples. It is validated for Flow Cytometry.Spezifikation
CD185 (CXCR5) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Blr1; Blr-1; Burkitt lymphoma receptor 1; burkitt lymphoma receptor 1 homolog; Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif) receptor 5); Burkitt lymphoma receptor 1, GTP-binding protein; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; CD185; CD185 antigen; chemochine (C-X-C motif) receptor 5; chemokine (C-X-C motif) receptor 5; CXC; C-X-C chemokine receptor type 5; CXC motif chemokine; C-X-C motif chemokine receptor 5; CXCR5; CXC-R5; CXCR-5; EGK_06966; Fusin; Gpcr6; GPR9; HUMSTR; LESTR; MDR15; MDR-15; Monocyte-derived receptor 15; neurolymphatic receptor; NLR | |
CXCR5 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
MU5UBEE | |
Flow Cytometry | |
Super Bright 436 | |
Mouse | |
Human, Non-human Primate, Rhesus Monkey | |
P32302 | |
643, 701792 | |
N-terminal extracellular domain | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CXCR5 |