Learn More
CD169 (Siglec-1) Monoclonal Antibody (SER-4), eFluor™ 660, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marke: Affymetrix eBioscience 50-5755-82
Additional Details : Gewicht : 0.09500kg
Beschreibung
Description: The SER-4 monoclonal antibody reacts with mouse CD169, which is also known as Sialoadhesin and Siglec-1. CD169 is a 185-kDa member of the Siglec family of adhesion and endocytic receptors that recognize sialylated glycans. This receptor is expressed on a subset of macrophages that are found with high frequency in the spleen, liver, lymph node, and bone marrow. CD169 can also be detected on monocytes in the bone marrow and periphery. This receptor plays a role in cell-cell adhesion, as well as internalization of sialylated virus and bacteria. This endocytic pathway allows CD169+ macrophages to present lipid antigens to iNKT cells, thereby leading to their activation. Studies have also demonstrated that direct interaction of CD169 with regulatory T cells inhibits their proliferation. The SER-4 antibody has been reported to have antagonist activity. Applications Reported: This SER-4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SER-4 antibody has been tested by flow cytometric anaylsis of mouse bone marrow cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
CD169 (Siglec-1) Monoclonal antibody specifically detects CD169 (Siglec-1) in Mouse samples. It is validated for Flow Cytometry.Spezifikation
CD169 (Siglec-1) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q62230 | |
SIGLEC1 | |
Affinity chromatography | |
RUO | |
20612 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
SER-4 | |
eFluor 660 | |
SIGLEC1 | |
Cd169; dJ1009E24.1; macrophage receptor; p210; pSn; SA; SER; sheep erythrocyte receptor; sialic acid binding Ig like lectin 1; sialic acid binding Ig-like lectin 1, sialoadhesin; Sialic acid-binding Ig-like lectin 1; sialic acid-binding immunoglobulin-like lectin 1; sialoadhesin; SIGLEC1; Siglec-1; SN | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.