Learn More
CD166 (ALCAM) Monoclonal Antibody (3A6), Super Bright™ 645, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
185.00 CHF - 388.00 CHF
Spezifikation
Antigen | CD166 (ALCAM) |
---|---|
Klon | 3A6 |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15983634
|
Affymetrix eBioscience
64-1668-41 |
25 Tests |
185.00 CHF
25 Stück |
Verfügbar ab: 26-04-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15993634
|
Affymetrix eBioscience
64-1668-42 |
100 Tests |
388.00 CHF
100 Stück |
Verfügbar ab: 26-04-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Description: This 1RNM44N monoclonal antibody recognizes mouse CD170, also known as Siglec F. Siglec F is a cell surface lectin belonging to the Ig superfamily that binds glycoconjugates containing sialic acids that are commonly found on various cell types. The cytoplasmic domain of Siglec F contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) that initiates intracellular signaling upon ligand binding. Siglec F is expressed mostly on eosinophils and alveolar macrophages and lower levels of this receptor have also been reported on immature myelomonocytic cells. Mouse Siglec F is a functional ortholog of human Siglec 8, however, unlike human Siglec 8, mouse Siglec F is not expressed on mast cells. This 1RNM44N antibody will recognize a formaldehyde-fixed epitope. Applications Reported: This 1RNM44N antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1RNM44N antibody has been tested by flow cytometric analysis of thioglycolate-elicited peritoneal macrophages. This may be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
CD166 (ALCAM) Monoclonal antibody specifically detects CD166 (ALCAM) in Human samples. It is validated for Flow Cytometry.Spezifikation
CD166 (ALCAM) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product | |
ALCAM | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ALCAM |
3A6 | |
Flow Cytometry | |
Super Bright 645 | |
Mouse | |
Human | |
Q13740 | |
214 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |