Learn More
CD16 Monoclonal Antibody (eBioCB16 (CB16)), Super Bright™ 645, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen 64-0168-41
Additional Details : Gewicht : 0.01000kg
Beschreibung
Description: The eBio1D3 monoclonal antibody reacts with mouse CD19, a 95 kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, MHC class II, and CD19 form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells. Applications Reported: This eBio1D3(1D3) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio1D3(1D3) antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No.
It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product No. SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 645 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. CD16 (FCGR3A) is a 50-65 kDa cell surface molecule that exists in two forms - a transmembranous form expressed by NK cells and some T cells, and a phosphatidylinositol linked form expressed by granulocytes. CD16 is a low affinity receptor for IgG (FcR III), and is an important receptor mediating ADCC by NK cells. Human CD16 is expressed in two forms – FCGR3A and FCGR3B. FCGR3A is associated with the FcepsilonRI-gamma subunit and is responsible for antibody-dependent NK cell cytotoxicity. Mast cell FCGR3A is associated, with FcepsilonRI-beta subunit. Besides IgG, FCGR3A can be triggered also by oligomeric IgE. FCGR3B is a GPI-linked monomeric receptor expressed on neutrophils and is involved in their activation and induction of a pro-adhesive phenotype. Diseases associated with CD16 dysfunction include immunodeficiency 20 and systemic lupus erythematosus.Spezifikation
CD16 | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
O75015, P08637 | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
eBioCB16 (CB16) | |
Super Bright 645 | |
FCGR3A | |
FCGR3A, FCGR3B | |
Affinity chromatography | |
RUO | |
2214, 2215 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |